missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxyphenylglycolaldehyde (Dopegal), TRC

Product Code. 30006198
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006198 5 mg 5mg
30012349 1 mg 1mg
30007920 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006198 Supplier Toronto Research Chemicals Supplier No. TRCD4546555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13023-73-9
Molecular Formula C8 H8 O4
Molecular Weight (g/mol) 168.15
Synonym 3,4-Dihydroxyphenylglycolic Aldehyde;
IUPAC Name 2-(3,4-dihydroxyphenyl)-2-hydroxyacetaldehyde
SMILES OC(C=O)c1ccc(O)c(O)c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C8H8O4/c9-4-8(12)5-1-2-6(10)7(11)3-5/h1-4,8,10-12H
Formula Weight 168.0423
Chemical Name or Material 3,4-Dihydroxyphenylglycolaldehyde (Dopegal)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.