missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzylamine Hydrobromide, MP Biomedicals™

Product Code. 11473112 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473112 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473112 Supplier MP Biomedicals Supplier No. 0215374083
  • ONLINE EXCLUSIVE PRICE
    47.40 EUR
    45.15 EUR / 250mg
    Save 2.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16290-26-9
Molecular Formula C7H10BrNO2
MDL Number MFCD00012859
Quantity 250 mg
Synonym 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, dhba hydrobromide, acmc-1afa4, 1, 4-aminomethyl-, hydrobromide, 4-aminomethyl catechol hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide
InChI Key BVFZTXFCZAXSHN-UHFFFAOYSA-N
SMILES [H+].[Br-].NCC1=CC=C(O)C(O)=C1
IUPAC Name hydrogen 4-(aminomethyl)benzene-1,2-diol bromide
Molecular Weight (g/mol) 220.07
PubChem CID 13343562
Formula Weight 220.1
Physical Form Solid
Chemical Name or Material 3, 4-Dihydroxybenzylamine Hydrobromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.