Learn More
3,4-Dihydro-2H-pyran, Sigma-Aldrich™
Product Code. 15888890
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15888890 250 mL 250mL
15878890 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15888890 Supplier Sigma-Aldrich Supplier No. 8029710250
 Restricted Item
Estimated Shipment: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-87-2
Grade Research
Molecular Weight (g/mol) 84.12
Specifications
Melting Point -70°C
Density 0.929 g/cm3 at 20°C
Boiling Point 85.5°C/1013 hPa
MDL Number MFCD00006558
Quantity 250 mL
Synonym 3,4-Dihydro-2H-pyran, 3,4-Dihydro-2H-pyran