Learn More
3,4-Dihydro-2H-1,4-benzoxazine, 97%
Product Code. p-7032246
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426487 1 g 1g
15436487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15426487 Supplier Thermo Scientific Alfa Aesar Supplier No. H33641.03
Add to basket
CAS: 5735-53-5 | C8H9NO | 135.166 g/mol
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5735-53-5
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD02181098
InChI Key YRLORWPBJZEGBX-UHFFFAOYSA-N
Synonym benzomorpholine, 3,4-dihydro-2h-benzo b 1,4 oxazine, 3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro, 2,3-dihydro-1,4-benzoxazine, akos bc-1149, 3,4-dihydro-2h-1,4-benzoxazin, pubchem22677, acmc-1amxt, 3,4-dihydro-2h-1,4-benzooxazine
PubChem CID 585096
IUPAC Name 3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=CC=CC=C2N1
Specifications
Boiling Point 85°C (1 mmHg)
Quantity 1 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 3,4-Dihydro-2H-1,4-benzoxazine