missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dicaffeoylquinic acid, MedChemExpress

Product Code. p-200098835
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471288 5 mg 5mg
16468848 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471288 Supplier Medchem Express Supplier No. HYN0057
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 3,4-Dicaffeoylquinic acid (3,4-Di-O-caffeoylquinic acid), naturally isolated from Laggera alata, has antioxidative, DNA protective, neuroprotective and hepatoprotective properties. 3,4-Dicaffeoylquinic acid exerts apoptosis-mediated cytotoxicity and α-glucosidase inhibitory effects. 3,4-Dicaffeoylquinic acid possesses a unique mechanism of anti-influenza viral activity, that is, enhancing viral clearance by increasing TRAIL.

3,4-Dicaffeoylquinic acid (3,4-Di-O-caffeoylquinic acid), naturally isolated from Laggera alata, has antioxidative, DNA protective, neuroprotective and hepatoprotective properties. 3,4-Dicaffeoylquinic acid exerts apoptosis-mediated cytotoxicity and α-glucosidase inhibitory effects. 3,4-Dicaffeoylquinic acid possesses a unique mechanism of anti-influenza viral activity, that is, enhancing viral clearance by increasing TRAIL.

Specifications

Chemical Name or Material 3,4-Dicaffeoylquinic acid
Physical Form Solid
CAS 14534-61-3
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H24O12
Synonym 3,4-Di-O-caffeoylquinic acid Isochlorogenic acid B
Solubility Information DMSO : 50 mg/mL (96.81 mM; Need ultrasonic)
SMILES O=C([C@@]1(O)C[C@@H](OC(/C=C/C2=CC=C(O)C(O)=C2)=O)[C@H](OC(/C=C/C3=CC=C(O)C(O)=C3)=O)[C@H](O)C1)O
Molecular Weight (g/mol) 516.45
Formula Weight 516.45
Percent Purity 98.06%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.