Learn More
3’,4’-Dibenzyloxy-1-phenyl-2-butanone, TRC
Specifications
Specifications
| Chemical Name or Material | 3',4'-Dibenzyloxy-1-phenyl-2-butanone |
| CAS | 24538-59-8 |
| Molecular Formula | C24 H24 O3 |
| Quantity | 50 mg |
| Synonym | 1-[3,4-Bis(phenylmethoxy)phenyl]-1-butanone |
| InChI Formula | InChI=1S/C24H24O3/c1-2-9-22(25)21-14-15-23(26-17-19-10-5-3-6-11-19)24(16-21)27-18-20-12-7-4-8-13-20/h3-8,10-16H,2,9,17-18H2,1H3 |
| SMILES | CCCC(=O)c1ccc(OCc2ccccc2)c(OCc3ccccc3)c1 |
| IUPAC Name | 1-[3,4-bis(phenylmethoxy)phenyl]butan-1-one |
| Molecular Weight (g/mol) | 360.446 |
| Formula Weight | 360.173 |