missing translation for 'onlineSavingsMsg'
Learn More

3-[(4-Chlorophenylsulfonyl)methyl]benzamidoxime, Alfa Aesar™

Product Code. 11460150
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460150 5 g 5g
11450150 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460150 Supplier Thermo Scientific Alfa Aesar Supplier No. H50633.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 583059-52-3
Molecular Formula C14H13ClN2O3S
Molecular Weight (g/mol) 324.779
MDL Number MFCD08060949
InChI Key JEBLYWBMDYPQRL-UHFFFAOYSA-N
Synonym chembl95609, 3-4-chlorophenyl sulfonyl methyl-n-hydroxybenzimidamide, 3-4-chlorophenyl sulfonyl methyl-n-hydroxybenzenecarboximidamide, 3-4-chlorophenylsulfonyl methyl benzamidoxime, 3-4-chlorophenyl sulfonyl methyl-n'-hydroxybenzenecarboximidamide, 3-4-chloro-benzenesulfonylmethyl-n-hydroxy-benzamidine, 4-chloro-1-3-hydroxyamino iminomethyl phenyl methyl sulfonyl benzene
PubChem CID 9584159
IUPAC Name 3-[(4-chlorophenyl)sulfonylmethyl]-N'-hydroxybenzenecarboximidamide
SMILES C1=CC(=CC(=C1)CS(=O)(=O)C2=CC=C(C=C2)Cl)C(=NO)N

Specifications

Quantity 5 g
Formula Weight 324.79
Chemical Name or Material 3-[(4-Chlorophenylsulfonyl)methyl]benzamidoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.