missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',4'-Anhydro Vincristine Ditartrate, TRC
Specifications
Specifications
| Chemical Name or Material | 3',4'-Anhydro Vincristine Ditartrate |
| Synonym | 3',4'-Didehydro-4'-deoxy-22-oxo-vincaleukoblastine Ditartrate∣3',4'-Anhydroleurocristine Ditartrate∣3',4'-Anhydrovincristine Ditartrate∣Anhydroleurocristine Ditartrate∣Anhydrovincristine Ditartrate∣ |
| Quantity | 2.5 mg |
| Shipping Condition | Room Temperature |
| Molecular Formula | C46 H54 N4 O9 . C4 H6 O6 |
| Formula Weight | 956.406 |
| Molecular Weight (g/mol) | 957.029 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?