Learn More
3-(4-Aminophenyl)acrylic acid, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 2393-18-2 |
| Molecular Formula | C9H9NO2 |
| MDL Number | MFCD00017117 |
| Quantity | 25 g |
| SMILES | O=C(O)/C=C/C1=CC=C(N)C=C1 |
| Molecular Weight (g/mol) | 163.17 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-(4-Aminophenyl)acrylic acid |