missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4,5-Dimethylthiazolyl-2)-2,5-Diphenyl Tetrazolium Bromide, MP Biomedicals
Shop All MP Biomedicals ProductsDescription
- Thiazolyl Blue Tetrazolium Blue (MTT) may be used in measurement of cell proliferation
- MTT produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells
- The blue crystals are solubilized with acidified isopropanol and the intensity is measured colorimetrically at 570 nm
- MTT has been used as a histochemical/cytochemical reagent and for the detection of NAD
- ADP-linked enzyme systems in tissue cannot be detected with MTT, due to binding of the cation by the cyanide trap used
- MTT is rapidly reduced to the formazan, which chelates with nickel, copper, and cobalt; the cobalt chelate has been used in oxidative systems
Specifications
Specifications
| Melting Point | 189°C to 199°C |
| Color | Yellow |
| Linear Formula | C18H16BrN5S |
| Quantity | 100 mg |
| Solubility Information | Soluble in methyl alcohol (heat), methanol (10mg/mL with heat - clear, yellow solution)and water (5mg/mL-clear to very slightly hazy, yellow to bright yellow solution). |
| Formula Weight | 414.3 |
| Percent Purity | ∼98% |
| Physical Form | Powder |
| Chemical Name or Material | 3-(4,5-DIMETHYLTHIAZOLYL-2)-2,5-DIPHENYL TETRAZOLIUM BROMIDE |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?