Learn More
3-[4-(3-Chlorophenyl)piperazin-1-yl]propyl Isobutyl Ether Hydrochloride, Mikromol™
Description
3-[4-(3-Chlorophenyl)piperazin-1-yl]propyl Isobutyl Ether Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 3-[4-(3-Chlorophenyl)piperazin-1-yl]propyl Isobutyl Ether Hydrochloride |
| CAS | 2470441-00-8 |
| Analyte or Component Names | 3-[4-(3-Chlorophenyl)piperazin-1-yl]propyl Isobutyl Ether Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H27 Cl N2 O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 3-[4-(3-Chlorophenyl)piperazin-1-yl]propyl Isobutyl Ether Hydrochloride, Trazodone Hydrochloride Imp. G (BP) as Hydrochloride, Trazodone Hydrochloride Impurity G, Trazodone Impurity G |
| InChI Formula | InChI=1S/C17H27ClN2O.ClH/c1-15(2)14-21-12-4-7-19-8-10-20(11-9-19)17-6-3-5-16(18)13-17;/h3,5-6,13,15H,4,7-12,14H2,1-2H3;1H |
| SMILES | Cl.CC(C)COCCCN1CCN(CC1)c2cccc(Cl)c2 |
| IUPAC Name | 1-(3-chlorophenyl)-4-[3-(2-methylpropoxy)propyl]piperazine;hydrochloride |
| Show More |