Learn More
3-[3-(Trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™
Product Code. 15454767
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454767 5 g 5g
15444767 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15454767 Supplier Thermo Scientific Alfa Aesar Supplier No. H32088.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 585-50-2
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.18
MDL Number MFCD00090487
InChI Key YLTJJMIWCCJIHI-UHFFFAOYSA-N
Synonym 3-3-trifluoromethyl phenyl propanoic acid, 3-3-trifluoromethylphenyl propionic acid, benzenepropanoic acid, 3-trifluoromethyl, 3-trifluoromethyl hydrocinnamic acid, unii-4u44xhh79h, 3-3'-trifluoromethylphenyl-propionic acid, 3-3-trifluoromethyl phenyl propionic acid, cinacalcet m5, m-trifluoromethyl hydrocinnamic acid
PubChem CID 2777683
IUPAC Name 3-[3-(trifluoromethyl)phenyl]propanoic acid
SMILES OC(=O)CCC1=CC(=CC=C1)C(F)(F)F
Specifications
Melting Point 34°C to 37°C
Quantity 5 g
Formula Weight 218.17
Percent Purity 97%
Chemical Name or Material 3-[3-(Trifluoromethyl)phenyl]propionic acid