Learn More
3,3-Dimethyl-1-(3-nitrophenyl-2,4,5,6-d4)triaz-1-ene, TRC
Product Code. 30179759
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179759 25 mg 25mg
30179899 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30179759 Supplier Toronto Research Chemicals Supplier No. TRCD15346225MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2519481-67-3
Molecular Formula C8 D4 H6 N4 O2
Molecular Weight (g/mol) 198.215
Synonym 1-(m-Nitrophenyl)-3,3-dimethyltriazine-d4, 3,3-Dimethyl-1-(3-nitrophenyl)triazene-d4, NSC 82309-d4
IUPAC Name N-methyl-N-[(E)-(2,3,4,6-tetradeuterio-5-nitrophenyl)diazenyl]methanamine
SMILES [2H]c1c([2H])c(\N=N\N(C)C)c([2H])c(c1[2H])N(=O)=O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C8H10N4O2/c1-11(2)10-9-7-4-3-5-8(6-7)12(13)14/h3-6H,1-2H3/b10-9+/i3D,4D,5D,6D
Formula Weight 198.105
Chemical Name or Material 3,3-Dimethyl-1-(3-Nitrophenyl-2,4,5,6-D4)Triaz-1-Ene