missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,3-Dimethoxy-2-butanone, 98%

Product Code. 15441529
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15441529 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15441529 Supplier Alfa Aesar™ Supplier No. H64365.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 21983-72-2
Color Colorless to Yellow
Density 0.987g/mL
Boiling Point 145°C to 146°C
Molecular Formula C6H12O3
Refractive Index 1.4070
MDL Number MFCD00075266
Quantity 25 g
UN Number UN1224
Beilstein 1752768
Synonym 3,3-dimethoxy-2-butanone, 2-butanone, 3,3-dimethoxy, acmc-20aa6y, 3.3-dimethoxy-2-butanone, 3,3-dimethoxy-2-oxobutane, 3,3-dimethoxy-butan-2-one, ksc495e8l
InChI Key UFQBSPGKRRSATO-UHFFFAOYSA-N
SMILES COC(C)(OC)C(C)=O
IUPAC Name 3,3-dimethoxybutan-2-one
Molecular Weight (g/mol) 132.16
PubChem CID 140871
Formula Weight 132.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3,3-Dimethoxy-2-butanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.