missing translation for 'onlineSavingsMsg'
Learn More

3,3',5-Triiodo Thyroacetic Acid-13C6, TRC

Product Code. 30456075
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456075 1 mg 1mg
30454204 5 mg 5mg
30454619 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456075 Supplier Toronto Research Chemicals Supplier No. TRCT7953221MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,3',5-Triiodo Thyroacetic Acid-13C6
Molecular Formula 13C6 C8 H9 I3 O4
Quantity 1 mg
Synonym 4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodobenzeneacetic Acid-13C6, Nidolin-13C6, T3A-13C6, Tiratricol-13C6, Triac-13C6, Triacana-13C6, [(4-Hydroxy-3-iodo-4-phenoxy)-3,5-diiodophenyl]acetic Acid-13C6
InChI Formula InChI=1S/C14H9I3O4/c15-9-6-8(1-2-12(9)18)21-14-10(16)3-7(4-11(14)17)5-13(19)20/h1-4,6,18H,5H2,(H,19,20)/i1+1,2+1,6+1,8+1,9+1,12+1
SMILES O[13c]1[13cH][13cH][13c](Oc2c(I)cc(CC(=O)O)cc2I)[13cH][13c]1I
IUPAC Name 2-[4-(4-hydroxy-3-iodo(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-yl)oxy-3,5-diiodophenyl]acetic acid
Molecular Weight (g/mol) 627.888
Formula Weight 627.784
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.