Learn More
3,3,4,4,4-Pentafluorobutylamine, TRC
Product Code. 30011555
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011555 500 mg 500mg
30011221 50 mg 50mg
30010435 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011555 Supplier Toronto Research Chemicals Supplier No. TRCP227543500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 139978-28-2
Molecular Formula C4 H6 N F5
Molecular Weight (g/mol) 163.09
Synonym 3,3,4,4,4-Pentafluoro-1-butanamine;
IUPAC Name 3,3,4,4,4-pentafluorobutan-1-amine
SMILES NCCC(F)(F)C(F)(F)F
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C4H6F5N/c5-3(6,1-2-10)4(7,8)9/h1-2,10H2
Formula Weight 163.042
Chemical Name or Material 3,3,4,4,4-Pentafluorobutylamine