missing translation for 'onlineSavingsMsg'
Learn More

3,3,4,4,4-Pentafluorobutylamine, TRC

Product Code. 30011221
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011221 50 mg 50mg
30010435 100 mg 100mg
30011555 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011221 Supplier Toronto Research Chemicals Supplier No. TRCP22754350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139978-28-2
Molecular Formula C4 H6 N F5
Molecular Weight (g/mol) 163.09
Synonym 3,3,4,4,4-Pentafluoro-1-butanamine;
IUPAC Name 3,3,4,4,4-pentafluorobutan-1-amine
SMILES NCCC(F)(F)C(F)(F)F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C4H6F5N/c5-3(6,1-2-10)4(7,8)9/h1-2,10H2
Formula Weight 163.042
Chemical Name or Material 3,3,4,4,4-Pentafluorobutylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.