missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Methoxyphenoxy)piperidine

Product Code. 11469760
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469760 1 g 1g
11459760 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469760 Supplier Alfa Aesar™ Supplier No. H50988.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 902837-27-8
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD08061100
InChI Key QWVSMVKBNRCNFJ-UHFFFAOYSA-N
Synonym 3-2-methoxyphenoxy piperidine, piperidine, 3-2-methoxyphenoxy, pubchem18383
PubChem CID 24902299
IUPAC Name 3-(2-methoxyphenoxy)piperidine
SMILES COC1=CC=CC=C1OC2CCCNC2

Specifications

Quantity 1 g
Formula Weight 207.27
Chemical Name or Material 3-(2-Methoxyphenoxy)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.