missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Isopropylphenoxy)piperidine, Thermo Scientific™

Product Code. 11419780
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419780 250 mg 250mg
11429780 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419780 Supplier Thermo Scientific Alfa Aesar Supplier No. H50996.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902837-35-8
Molecular Formula C14H21NO
Molecular Weight (g/mol) 219.328
MDL Number MFCD08061106
InChI Key DFJMRKMGDKDNAX-UHFFFAOYSA-N
Synonym 3-2-isopropylphenoxy piperidine, 3-2-propan-2-yl phenoxy piperidine
PubChem CID 44721294
IUPAC Name 3-(2-propan-2-ylphenoxy)piperidine
SMILES CC(C)C1=CC=CC=C1OC2CCCNC2

Specifications

Quantity 250 mg
Formula Weight 219.33
Chemical Name or Material 3-(2-Isopropylphenoxy)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.