missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethyl)-1-Boc-azetidine, 95%

Product Code. 15456938
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456938 250 mg 250mg
15446938 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456938 Supplier Thermo Scientific Alfa Aesar Supplier No. H60469.MD
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 898271-20-0 | C10H20N2O2 | 200.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 898271-20-0
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD09878606
InChI Key XQNQFOKZUQWWDO-UHFFFAOYSA-N
Synonym 1-boc-3-aminoethylazetidine, tert-butyl 3-2-aminoethyl azetidine-1-carboxylate, 1-boc-3-aminoethyl azetidine, 1-n-boc-3-aminoethyl azetidine, 1-boc-3-2-aminoethyl azetidine, 3-2-aminoethyl-1-boc-azetidine, 1-azetidinecarboxylicacid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-azetidinecarboxylic acid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, tert-butyl 3-2-aminoethyl-1-azetidinecarboxylate
PubChem CID 44630603
IUPAC Name tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)CCN

Specifications

Quantity 250 mg
Formula Weight 200.28
Percent Purity 95%
Chemical Name or Material 3-(2-Aminoethyl)-1-Boc-azetidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.