Learn More
3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarbonyl Chloride, TRC
Product Code. 30384918
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384918 1 g
30378712 250 mg
30381526 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30384918 Supplier Toronto Research Chemicals Supplier No. TRCD4356781G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 52314-67-7
Molecular Formula C8H9Cl3O
Molecular Weight (g/mol) 227.52
Synonym 3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarbonyl Chloride, 2,2-Dimethyl-3-(2,2-dichloroethenyl)cyclopropanecarbonyl Chloride, 2,2-Dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarbonyl Chloride, 3-(2,2-Dichlorovinyl)-2,2-dimethyl-1-cyclopropanecarbonyl Chloride
IUPAC Name 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carbonyl chloride
SMILES CC1(C)C(C=C(Cl)Cl)C1C(=O)Cl
Specifications
Quantity 1 g
InChI Formula InChI=1S/C8H9Cl3O/c1-8(2)4(3-5(9)10)6(8)7(11)12/h3-4,6H,1-2H3
Formula Weight 225.9719
Chemical Name or Material 3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarbonyl Chloride