missing translation for 'onlineSavingsMsg'
Learn More

3,11-Dimethylchrysene, TRC

Product Code. 30010927
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
Unit Size:
10mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010927 10 mg 10mg
30005417 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010927 Supplier Toronto Research Chemicals Supplier No. TRCD46494010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139493-40-6
Molecular Formula C20H16
Molecular Weight (g/mol) 256.34
SMILES CC1=CC2=CC=C(C)C=C2C3=C1C(C=CC=C4)=C4C=C3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H16/c1-13-7-8-16-12-14(2)20-17-6-4-3-5-15(17)9-10-18(20)19(16)11-13/h3-12H,1-2H3
Chemical Name or Material 3,11-Dimethylchrysene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.