Learn More
3-(1,3-Dithian-2-ylidene)pentane-2,4-dione, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(1,3-Dithian-2-ylidene)-pentane-2,4-dione |
| CAS | 55727-23-6 |
| Molecular Formula | C9 H12 O2 S2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C9H12O2S2/c1-6(10)8(7(2)11)9-12-4-3-5-13-9/h3-5H2,1-2H3 |
| SMILES | CC(=O)C(=C1SCCCS1)C(=O)C |
| IUPAC Name | 3-(1,3-dithian-2-ylidene)pentane-2,4-dione |
| Molecular Weight (g/mol) | 216.32 |
| Formula Weight | 216.0279 |