Learn More
(2S,4R)-1-(2-Aminoacetyl)-4-hydroxypyrrolidine-2-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (2S,4R)-1-(2-Aminoacetyl)-4-hydroxypyrrolidine-2-carboxylic Acid |
| CAS | 24587-32-4 |
| Molecular Formula | C7H12N2O4 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C7H12N2O4/c8-2-6(11)9-3-4(10)1-5(9)7(12)13/h4-5,10H,1-3,8H2,(H,12,13)/t4-,5+/m1/s1 |
| SMILES | NCC(=O)N1C[C@H](O)C[C@H]1C(=O)O |
| IUPAC Name | (2S,4R)-1-(2-aminoacetyl)-4-hydroxypyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 188.18 |
| Formula Weight | 188.0797 |