Learn More
(-)-(2S,3S)-Viridifloric Acid Ethyl Ester-d7, TRC
Specifications
Specifications
| Chemical Name or Material | (-)-(2S,3S)-Viridifloric Acid Ethyl Ester-d7 |
| Molecular Formula | C9 D7 H11 O4 |
| Quantity | 2.5 mg |
| Synonym | (-)-Viridifloric Acid Ethyl Ester-d7, Ethyl (-)-Viridiflorate-d7, [S-(R*,R*)]-2,3-Dihydroxy-2-(1-methylethyl)-butanoic Acid Ethyl Ester-d7, (2S,3S)-2,3-Dihydroxy-2-(1-methylethyl)butanoic Acid Ethyl Ester-d7 |
| InChI Formula | InChI=1S/C9H18O4/c1-5-13-8(11)9(12,6(2)3)7(4)10/h6-7,10,12H,5H2,1-4H3/t7-,9-/m1/s1/i2D3,3D3,6D |
| SMILES | [2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])[C@@](O)([C@@H](C)O)C(=O)OCC |
| IUPAC Name | ethyl (2R)-3,4,4,4-tetradeuterio-2-hydroxy-2-[(1R)-1-hydroxyethyl]-3-(trideuteriomethyl)butanoate |
| Molecular Weight (g/mol) | 197.28 |
| Formula Weight | 197.164 |