Learn More
(2S,3S)-3-Hydroxynorleucine, TRC
Product Code. 30396110
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30396110 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30396110 Supplier Toronto Research Chemicals Supplier No. TRCH9497551G
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2S,3S)-3-Hydroxynorleucine
CAS 10148-68-2
Molecular Formula C6H13NO3
Quantity 1 g
Synonym erythro-3-Hydroxy-norleucine, (2S,3S)-2-Amino-3-hydroxy-hexanoic Acid
InChI Formula InChI=1S/C6H13NO3/c1-2-3-4(8)5(7)6(9)10/h4-5,8H,2-3,7H2,1H3,(H,9,10)/t4-,5-/m0/s1
SMILES CCC[C@H](O)[C@H](N)C(=O)O
IUPAC Name (2S,3S)-2-amino-3-hydroxyhexanoic acid
Molecular Weight (g/mol) 147.17
Formula Weight 147.0895