Learn More
(2S,3R,4E)-2-Amino-4-tetradecene-1,3-diol, TRC
Specifications
Specifications
| Chemical Name or Material | (2S,3R,4E)-2-Amino-4-tetradecene-1,3-diol |
| CAS | 24558-60-9 |
| Molecular Formula | C14H29NO2 |
| Quantity | 25 mg |
| Synonym | (E)-D-erythro-2-Amino-4-tetradecene-1,3-diol, C14-Sphingosine, Tetradecasphing-4-enine |
| InChI Formula | InChI=1S/C14H29NO2/c1-2-3-4-5-6-7-8-9-10-11-14(17)13(15)12-16/h10-11,13-14,16-17H,2-9,12,15H2,1H3/b11-10+/t13-,14+/m0/s1 |
| SMILES | CCCCCCCCC\C=C\[C@@H](O)[C@@H](N)CO |
| IUPAC Name | (E,2S,3R)-2-aminotetradec-4-ene-1,3-diol |
| Molecular Weight (g/mol) | 243.39 |
| Formula Weight | 243.2198 |