missing translation for 'onlineSavingsMsg'
Learn More

(2S,2'S)-1,1'-[[(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)imino]bis(1-oxo-2,1-ethanediyl)]bis-2-pyrrolidinecarbonitrile, TRC

Product Code. 17992905
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992905 500 mg 500mg
17942255 100 mg 100mg
17953215 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992905 Supplier Toronto Research Chemicals Supplier No. TRCH971385500MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1036959-23-5
Molecular Formula C24 H33 N5 O3
Molecular Weight (g/mol) 439.55
Synonym 2-Pyrrolidinecarbonitrile, 1,1'-[[(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)imino]bis(1-oxo-2,1-ethanediyl)]bis-, (2S,2'S)-, (2S,2'S)-1,1'-[[(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)imino]bis(1-oxo-2,1-ethanediyl)]bis[2-pyrrolidinecarbonitrile], (2S)-1-[[[2[(2S)-2-Cyanopyrrolidin-1-yl]-2-oxoethyl][(3-hydroxytricyclo[3.3.1.1(3,7)]dec-1-yl)-amino]]acetyl]-pyrrolidine-2-carbonitrile
IUPAC Name (2S)-1-[2-[[2-[(2S)-2-cyanopyrrolidin-1-yl]-2-oxoethyl]-(3-hydroxy-1-adamantyl)amino]acetyl]pyrrolidine-2-carbonitrile
SMILES OC12CC3CC(C1)CC(C3)(C2)N(CC(=O)N4CCC[C@H]4C#N)CC(=O)N5CCC[C@H]5C#N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C24H33N5O3/c25-12-19-3-1-5-28(19)21(30)14-27(15-22(31)29-6-2-4-20(29)13-26)23-8-17-7-18(9-23)11-24(32,10-17)16-23/h17-20,32H,1-11,14-16H2/t17?,18?,19-,20-,23?,24?/m0/s1
Formula Weight 439.2583
Percent Purity >95%
Chemical Name or Material (2S)-1-[2-[[2-((2S)-2-Cyanopyrrolidin-1-yl)-2-oxoethyl](3-hydroxy-1-adamantyl)amino]acetyl]pyrrolidine-2-carbonitrile
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.