Learn More
(2S)-2-Amino-4-methyl-1-[(2R)-2-methyloxiranyl]-1-pentanone Trifluoroacetate, TRC
Specifications
Specifications
| Chemical Name or Material | (2S)-2-Amino-4-methyl-1-[(2R)-2-methyloxiranyl]-1-pentanone Trifluoroacetate |
| CAS | 247068-85-5 |
| Molecular Formula | C9H17NO2 . C2HF3O2 |
| Quantity | 1 mg |
| Synonym | (2S)-2-Amino-4-methyl-1-[(2R)-2-methyl-2-oxiranyl]-1-pentanone 2,2,2-Trifluoroacetate |
| InChI Formula | InChI=1S/C9H17NO2.C2HF3O2/c1-6(2)4-7(10)8(11)9(3)5-12-9;3-2(4,5)1(6)7/h6-7H,4-5,10H2,1-3H3;(H,6,7)/t7-,9+;/m0./s1 |
| SMILES | CC(C)CC(N)C(=O)[C@@]1(C)CO1.OC(=O)C(F)(F)F |
| IUPAC Name | (2S)-2-amino-4-methyl-1-[(2R)-2-methyloxiran-2-yl]pentan-1-one;2,2,2-trifluoroacetic acid |
| Molecular Weight (g/mol) | 285.26 |