Learn More
(2RS)-(1-Hydroxycyclopentyl)(phenyl)acetic Acid, Mikromol™
Product Code. 30051719
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051719 25 mg 25mg
30052415 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30051719 Supplier Mikromol Supplier No. MM0580.010025
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-(1-Hydroxycyclopentyl)(phenyl)acetic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2RS)-(1-Hydroxycyclopentyl)(phenyl)acetic Acid
CAS 25209-52-3
Analyte or Component Names (2RS)-(1-Hydroxycyclopentyl)(phenyl)acetic Acid
Shipping Condition Room Temperature
Molecular Formula C13 H16 O3
Certifications/Compliance ISO 17025
Synonym Cyclopentolate Hydrochloride Imp. A (EP), Cyclopentolate Imp. A (EP), (2RS)-(1-Hydroxycyclopentyl)(phenyl)acetic Acid, Cyclopentolate Hydrochloride Impurity A, Cyclopentolate Impurity A
InChI Formula InChI=1S/C13H16O3/c14-12(15)11(10-6-2-1-3-7-10)13(16)8-4-5-9-13/h1-3,6-7,11,16H,4-5,8-9H2,(H,14,15)
SMILES OC(=O)C(c1ccccc1)C2(O)CCCC2
IUPAC Name 2-(1-hydroxycyclopentyl)-2-phenylacetic acid
Molecular Weight (g/mol) 220.26
Quantity 25 mg
Formula Weight 220.1099 g/mol