Learn More
(2RS)-1-(Ethylamino)-3-[[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy]propan-2-ol, Mikromol™
Product Code. 30050414
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050414 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30050414 Supplier Mikromol Supplier No. MM0440.150025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-1-(Ethylamino)-3-[[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy]propan-2-ol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2RS)-1-(Ethylamino)-3-[[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy]propan-2-ol
CAS 2469037-77-0
Analyte or Component Names (2RS)-1-(Ethylamino)-3-[[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy]propan-2-ol
Shipping Condition Room Temperature
Molecular Formula C11 H20 N4 O3 S
Certifications/Compliance ISO 9001
Synonym Timolol Maleate Imp I (EP): (2RS)-1-(Ethylamino)-3-[[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy]propan-2-ol
InChI Formula InChI=1S/C11H20N4O3S/c1-2-12-7-9(16)8-18-11-10(13-19-14-11)15-3-5-17-6-4-15/h9,12,16H,2-8H2,1H3
SMILES CCNCC(O)COc1nsnc1N2CCOCC2
IUPAC Name 1-(ethylamino)-3-[(4-morpholin-4-yl-1,2,5-thiadiazol-3-yl)oxy]propan-2-ol
Molecular Weight (g/mol) 288.366
Quantity 25 mg
Formula Weight 288.126 g/mol