Learn More
(2R,3S,4S,5R,6S)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, TRC
Specifications
Specifications
| Chemical Name or Material | (2R,3S,4S,5R,6S)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol |
| CAS | 2459303-35-4 |
| Molecular Formula | C21 H25 Cl O6 |
| Quantity | 50 mg |
| Synonym | ent-Dapagliflozin |
| InChI Formula | InChI=1S/C21H25ClO6/c1-2-27-15-6-3-12(4-7-15)9-14-10-13(5-8-16(14)22)21-20(26)19(25)18(24)17(11-23)28-21/h3-8,10,17-21,23-26H,2,9,11H2,1H3/t17-,18-,19+,20-,21+/m0/s1 |
| SMILES | CCOc1ccc(Cc2cc(ccc2Cl)[C@H]3O[C@@H](CO)[C@H](O)[C@@H](O)[C@@H]3O)cc1 |
| IUPAC Name | (2R,3S,4S,5R,6S)-2-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-6-(hydroxymethyl)oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 408.873 |
| Formula Weight | 408.134 |