Learn More
((2R,3S,4R,5R)-5-(3-Carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen phosphate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 1094-61-7 |
| Molecular Formula | C11H15N2O8P |
| MDL Number | MFCD00038748 |
| Quantity | 10 g |
| SMILES | O=P(OC[C@H]1O[C@@H]([N+]2=CC=CC(C(N)=O)=C2)[C@H](O)[C@@H]1O)([O-])O |
| Molecular Weight (g/mol) | 334.22 |
| Percent Purity | 97% |
| Chemical Name or Material | ((2R,3S,4R,5R)-5-(3-Carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen phosphate |