Learn More
2M Lithium Chloride Electrolyte, Electrode Filling Solution, Paragon Scientific™
Product Code. 16060200
Change view
Click to view available options
Quantity:
3 x 30 mL
5 x 30 mL
10 x 30 mL
Unit Size:
Pack of 10
Pack of 3
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16060200 3 x 30 mL Pack of 3
16070200 5 x 30 mL Pack of 5
16050200 10 x 30 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16060200 Supplier Paragon Scientific Supplier No. LCL303
 Restricted Item
Estimated Shipment: 22-12-2026
A high quality analytical reagent manufactured and certified compliant for use in ASTM D 664 / IP 177, the Standard Test Method for Acid Number of Petroleum products by Potentiometric Titration.
  • A Lithium Chloride Electrolyte Analytical Reagent manufactured to ASTM D 664 / IP 177.
  • Provided with a certificate which includes an estimation of uncertainty of manufacture.
  • Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
  • Convenient 30mL bottle size for direct filling into the electrode.
  • Multi pack option available
  • Cost effective and time saving
  • Certificate of Manufacture & MSDS included
  • 6 Month shelf life
Specifications
Quantity 3 x 30 mL
compliance-icons compliance-icons
Product Identifier
  • 2M Lithium Chloride Electrolyte, Electrode Filling Solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, lithium chloride