Learn More
(2E,6E)-3,7,11-Trimethyldodeca-2,6,10-trien-1-yl Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | (2E,6E)-3,7,11-Trimethyldodeca-2,6,10-trien-1-yl Acetate |
| CAS | 4128-17-0 |
| Molecular Formula | C17H28O2 |
| Quantity | 10 g |
| Synonym | (2E,6E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol 1-Acetate, (2E,6E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol Acetate, (E,E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol Acetate, (2E,6E)-Farnesyl Acetate, (E,E)-Farnesyl Acetate, 2-trans-6-trans-Farnesyl Acetate, Acetic Acid (2E,6E)-3,7,11-Trimethyl-2,6,10-dodecatrienyl Ester, Acetic Acid Farnesyl Ester, all-trans-Farnesyl Acetate, trans,trans-Farnesol Acetate, trans,trans-Farnesyl Acetate, trans-2-trans-6-Farnesyl Acetate |
| InChI Formula | InChI=1S/C17H28O2/c1-14(2)8-6-9-15(3)10-7-11-16(4)12-13-19-17(5)18/h8,10,12H,6-7,9,11,13H2,1-5H3/b15-10+,16-12+ |
| SMILES | CC(=CCC\C(=C\CC\C(=C\COC(=O)C)\C)\C)C |
| IUPAC Name | [(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl] acetate |
| Molecular Weight (g/mol) | 264.4 |
| Formula Weight | 264.2089 |