missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BAI1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marca: R&D Systems AF4969-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BAI1 Polyclonal specifically detects BAI1 in Human samples. It is validated for Neutralization.
Especificaciones
| BAI1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 575 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Neutralization | |
| LYOPH | |
| Neutralization 2-8 ug/mL | |
| O14514 | |
| BAI1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human BAI1 Ala31-Thr879 Accession # O14514 | |
| 25 μg | |
| Primary | |
| Detects human BAI1 in direct ELISAs. In direct ELISAs, less than 2% cross-reactivity with recombinant human BAI3 is observed. | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido