Learn More
Adenosine-1'-13C, TRC
Product Code. 30321935
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321935 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30321935 Supplier Toronto Research Chemicals Supplier No. TRCA28040325MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Adenosine-1'-13C
CAS 201996-55-6
Molecular Formula 13C C9 H13 N5 O4
Quantity 25 mg
Synonym 9-β-D-Ribofuranosyl-9H-purin-6-amine-1'-13C, 9-β-D-Ribofuranosyladenine-1'-13C, Adenine Riboside-1'-13C, Adenocard-1'-13C, Adenocor-1'-13C, Adenogesic-1'-13C, Adenoscan-1'-13C, Adrekar-1'-13C, Boniton-1'-13C, D-Adenosine-1'-13C, 1-(6-Amino-9H-purin-9-yl)-1-deoxy-β-D-ribofuranose-1'-13C, Myocol-1'-13C, NSC 7652-1'-13C
InChI Formula InChI=1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7-,10-/m1/s1/i10+1
SMILES Nc1ncnc2c1ncn2[13C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O
IUPAC Name (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)(213C)oxolane-3,4-diol
Molecular Weight (g/mol) 268.234
Formula Weight 268.1