missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trenbolone Acetate, Mikromol™
Description
Trenbolone Acetate
Specifications
Specifications
| Chemical Name or Material | Trenbolone Acetate |
| CAS | 10161-34-9 |
| Analyte or Component Names | Trenbolone Acetate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C20 H24 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Estra-4,9,11-trien-3-one, 17β-hydroxy-, acetate (7CI,8CI), (17β)-17-(Acetyloxy)estra-4,9,11-trien-3-one, 17β-Acetoxy-Δ4,9,11-estratrien-3-one, 17β-Acetoxyestra-4,9,11-trien-3-one, 17β-Hydroxyestra-4,9,11-trien-3-one 17-acetate, 3-Oxo-17β-acetoxyestra-4,9,11-triene, Component T-H, Component T-S, Finaplix, Finaplix H, Finaplix-S, RU 1697, Trenbolone acetate, Trienbolone acetate |
| InChI Formula | InChI=1S/C20H24O3/c1-12(21)23-19-8-7-18-17-5-3-13-11-14(22)4-6-15(13)16(17)9-10-20(18,19)2/h9-11,17-19H,3-8H2,1-2H3/t17-,18+,19+,20+/m1/s1 |
| SMILES | CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CCC4=C3C=C[C@]12C |
| IUPAC Name | [(8S,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?