missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-S-(2-hydroxyethyl)-L-cysteine Dicyclohexylammonium Salt, TRC

Product Code. 17994145
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994145 250 mg 250mg
17981875 25 mg 25mg
17964095 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994145 Supplier Toronto Research Chemicals Supplier No. TRCA178911250MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1331896-18-4
Molecular Formula C7 H13 N O4 S . C12 H23 N
Molecular Weight (g/mol) 388.56
Synonym L-N-Acetyl-3-[(2-hydroxyethyl)thio]alanine Dicyclohexylammonium Salt, S-(2-Hydroxyethyl)mercapturic Acid Dicyclohexylammonium Salt
IUPAC Name (2R)-2-acetamido-3-(2-hydroxyethylsulfanyl)propanoic acid;N-cyclohexylcyclohexanamine
SMILES CC(=O)N[C@@H](CSCCO)C(=O)O.C1CCC(CC1)NC2CCCCC2

Specifications

InChI Formula InChI=1S/C12H23N.C7H13NO4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(10)8-6(7(11)12)4-13-3-2-9/h11-13H,1-10H2;6,9H,2-4H2,1H3,(H,8,10)(H,11,12)/t;6-/m.0/s1
Quantity 250 mg
Formula Weight 388.2396
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-Acetyl-S-(2-hydroxyethyl)-L-cysteine Dicyclohexylammonium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.