Learn More
Hamilton™ 25 μL 1700 Series Gas Tight Syringe
Product Code. 17824392
Change view
Click to view available options
Needle Gauge:
22s G
Custom
No Needle Included
Needle Point Style:
Blunt
Cone
Custom
No Needle Included
Sharp, Bevel, Curved, Non-coring
Connection Type:
Cemented
Luer Lock
Removable
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Needle Point Style Connection Type unitSize
17824392 22s G Sharp, Bevel, Curved, Non-coring Cemented Each
17834312 22s G Blunt Cemented Each
17814392 22s G Cone Cemented Each
17824312 22s G Sharp, Bevel, Curved, Non-coring Removable Each
17854202 22s G Blunt Removable Each
17824402 Custom Custom Cemented Each
17824192 No Needle Included No Needle Included Luer Lock Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17824392 Supplier Hamilton™ Supplier No. CAL80230
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
  • This series of syringes are ideal for dispensing volumes up to 25 μL.
  • A variety of different terminations and needle options are available in this series.
Specifications
Autoclavable Not Autoclavable
Material PTFE
Needle Gauge 22s G
Diameter (English) Inner 0.029 in.
Diameter (English) Inner Needle 0.0059 in.
Diameter (Metric) Inner Needle 0.15 mm
Diameter (English) Outer Needle 0.0283 in.
Diameter (English) Outer 0.305 in.
Diameter (Metric) Inner 0.729 mm
Diameter (Metric) Outer 7.75 mm
Diameter (Metric) Outer Needle 0.72 mm
Length (English) Needle 2 in.
Product Type Gas Tight Syringe
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 1702 N
Min. Temperature (Metric) 10°C
Quantity 1
Volume (Metric) 25 μL
Length (Metric) Needle 51 mm
Needle Point Style Sharp, Bevel, Curved, Non-coring
Connection Type Cemented