missing translation for 'onlineSavingsMsg'
Learn More

25,26-Dihydroxyvitamin D3, MedChemExpress

Product Code. 16449397
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449397 5 mg 5mg
16441559 1 mg 1mg
16426858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16449397 Supplier Medchem Express Supplier No. HY15830

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 25,26-Dihydroxyvitamin D3(25,26-dihydroxycholecalciferol) is a metabolite of vitamin D3 with intestinal calcium transport activity.

25,26-Dihydroxyvitamin D3(25,26-dihydroxycholecalciferol) is a metabolite of vitamin D3 with intestinal calcium transport activity.

Specifications

Chemical Name or Material 25,26-Dihydroxyvitamin D3
CAS 29261-12-9
Color White
Purity Grade Notes Research
Molecular Formula C27H44O3
Quantity 5 mg
Synonym 25,26-Dihydroxycholecalciferol
Solubility Information DMSO : 100 mg/mL (240.02 mM; Need ultrasonic)
SMILES C[C@@H]([C@H]1CC[C@@]2([H])/C(CCC[C@]12C)=C/C=C3C(CC[C@H](O)C\3)=C)CCCC(O)(C)CO
Molecular Weight (g/mol) 416.64
Formula Weight 416.64
Percent Purity 98.08%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.