missing translation for 'onlineSavingsMsg'
Learn More

24 Bisphenol S, TRC

Product Code. 30138073
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138073 2.5 g 2.5g
30004550 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138073 Supplier Toronto Research Chemicals Supplier No. TRCB5196202.5G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5397-34-2
Molecular Formula C12 H10 O4 S
Molecular Weight (g/mol) 250.27
Synonym Phenol, 2,4'-sulfonyldi- (6CI,7CI,8CI), 2-[(4-Hydroxyphenyl)sulfonyl]phenol, 2,4'-Bisphenol sulfone, 2,4'-Dihydroxydiphenyl sulfone, 2,4'-Sulfonyldiphenol, 2-(4-Hydroxyphenylsulfonyl)phenol, 24 Bisphenol S, 24BS, 4,2'-Dihydroxydiphenyl sulfone, BPS 24, BPS 24C, NSC 2432
IUPAC Name 2-(4-hydroxyphenyl)sulfonylphenol
SMILES Oc1ccc(cc1)S(=O)(=O)c2ccccc2O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C12H10O4S/c13-9-5-7-10(8-6-9)17(15,16)12-4-2-1-3-11(12)14/h1-8,13-14H
Formula Weight 250.03
Chemical Name or Material 2,4-Bisphenol S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.