Learn More
(20S)-Dexamethasone Epimeric Glycolic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (20S)-Dexamethasone Epimeric Glycolic Acid |
| CAS | 247075-09-8 |
| Molecular Formula | C22 H29 F O6 |
| Quantity | 2.5 mg |
| Synonym | (11β,16α, 20S)-9-Fluoro-11,17,20-trihydroxy-16-methyl-3-oxo-pregna-1,4-dien-21-oic Acid |
| InChI Formula | InChI=1S/C22H29FO6/c1-11-8-15-14-5-4-12-9-13(24)6-7-19(12,2)21(14,23)16(25)10-20(15,3)22(11,29)17(26)18(27)28/h6-7,9,11,14-17,25-26,29H,4-5,8,10H2,1-3H3,(H,27,28)/t11-,14+,15+,16+,17-,19+,20+,21+,22+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)[C@H](O)C(=O)O |
| IUPAC Name | (2S)-2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-hydroxy-acetic acid |
| Molecular Weight (g/mol) | 408.46 |
| Formula Weight | 408.1948 |