Learn More
Fisherbrand™ 20mm Crimp Cap
Product Code. 11895010 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue Lacquered
Clear Lacquered
Gold Lacquered
Green Lacquered
Plain
Red Lacquered
Product Type:
Center Hole Crimp Cap
Center Tear-off Crimp Cap
Complete Tear-off Crimp Cap
Unit Size:
Pack of 1000
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type Unit Size
11895010 Clear Lacquered Complete Tear-off Crimp Cap Pack of 1000
11805020 Plain Center Hole Crimp Cap Pack of 1000
11885010 Plain Center Tear-off Crimp Cap Pack of 1000
12616765 Gold Lacquered Center Hole Crimp Cap Pack of 1000
12626765 Red Lacquered Center Hole Crimp Cap Pack of 1000
12606505 Blue Lacquered Center Hole Crimp Cap Pack of 1000
13401309 Blue Lacquered Center Tear-off Crimp Cap Pack of 1000
15440130 Red Lacquered Center Tear-off Crimp Cap Pack of 1000
12646765 Green Lacquered Center Hole Crimp Cap Pack of 1000
15440120 Green Lacquered Center Tear-off Crimp Cap Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11895010
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
Aluminum crimp closures provide a secure leak-resistant seal.
  • 20mm
  • aluminum Cap
  • Aluminum seals must be applied with a crimping tool
TRUSTED_SUSTAINABILITY
Specifications
Color Clear Lacquered
Diameter (Metric) Outer 20 mm
Material Aluminum
Quantity 100
Closure Size ND20
Product Type Complete Tear-off Crimp Cap