missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2-propanediol-d5 (10 ug/mL in methanol), TRC

Product Code. 30240718
Change view
Click to view available options
Quantity:
20 x 1 mL
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240718 20 x 1 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240718 Supplier Toronto Research Chemicals Supplier No. TRCKIT890020X1ML
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 342611-01-2
Molecular Formula C3 2H5 H2 Cl O2
Quantity 20 x 1 mL
Synonym 3-MCPD D5, 3-chloro-1,2-propane-1,1,2,3,3-d5-diol
InChI Formula InChI=1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2/i1D2,2D2,3D
SMILES [2H]C([2H])(O)C([2H])(O)C([2H])([2H])Cl
IUPAC Name 3-chloro-1,1,2,3,3-pentadeuteriopropane-1,2-diol
Molecular Weight (g/mol) 115.57
Formula Weight 115.0448
Chemical Name or Material 3-Chloro-1,2-propanediol D5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.