Learn More
KGW Isotherm™ 2000mL Blue Metal Dewar Carrying Flask with Handle
Product Code. 10370571
Change view
Click to view available options
Capacity (Metric):
2 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10370571 2 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10370571 Supplier KGW Isotherm™ Supplier No. 10613
  • ONLINE EXCLUSIVE PRICE
    269.94 EUR
    250.17 EUR / Pack of 1
    Save 19.77 EUR
    7% Off
Estimated Shipment: 14-09-2026
Specifications
Diameter (Metric) Outer 120mm
For Use With (Application) Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants
Height (Metric) Inner 290mm
Height (Metric) 345mm
Includes Side Grip
Capacity (Metric) 2 L
Diameter (Metric) Inner 100 mm
Material Borosilicate Glass