Learn More
Fisherbrand™ 20-Well Tube Rack
Product Code. 11784016 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
Microtubes (0.5 to 2 mL)
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
11784016 Microtubes (0.5 to 2 mL) Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11784016
  • ONLINE EXCLUSIVE PRICE
    34.80 EUR
    30.22 EUR / Pack of 6
    Save 4.58 EUR
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
20-Well Tube Rack with interlocking racks for increased tube storage.
  • Interlock multiple racks to increase tube storage
  • Racks lock together on either side
  • Easily remove tubes using the hold-it finger slot on the front of the rack
  • Large molded coordinates for easy tube identification
  • Bi-level rack enables easy tube label identification
Specifications
Autoclavable Autoclavable
Color Blue
Holds Microtubes (0.5 to 2 mL)
Material Polypropylene
Positions 20 Wells
Shape Triangle
Stackable No
Sterility Non-sterile
For Use With (Equipment) 1.0 mL and 1.5/2.0 mL Tubes
Quantity 6 Pk.
Size 213 x 90 x 50 mm
Dimensions (L x W x H) 213 x 90 x 50 mm
Length (Metric) 21.1 cm
Length (English) 8.3 in.
Width (English) 3.5 in.
Height (English) 2 in.
Product Type Microtube Rack
Shelves None
Height (Metric) 5 cm
Array 2 x 10
Width (Metric) 9 cm