Learn More
Fisherbrand™ Two-Way Swivel Scaffolding Connectors
Product Code. 11558112 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Product Type:
2-Way Connector
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11558112 2-Way Connector Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11558112
Estimated Shipment: 04-02-2027
Add to basket
Add to basket
Used for fixing two rods or tubes at any angle to each other. Fisherbrand™ Two-Way Swivel Scaffolding Connectors can be tightened by hand.
  • With thumbscrews
  • The two components are restrained from swiveling freely by a tension washer
Specifications
Product Type 2-Way Connector
For Use With (Application) Swivel 2-Way