Learn More
2-(Trimethylacetyl)thiophene, 98%
Product Code. 11351947
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351947 5 g 5g
11361947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11351947 Supplier Thermo Scientific Alfa Aesar Supplier No. L07560.06
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
CAS: 20409-48-7 | C9H12OS | 168.254 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20409-48-7
Molecular Formula C9H12OS
Molecular Weight (g/mol) 168.254
MDL Number MFCD00051668
InChI Key PMUOKYKJIKZPIR-UHFFFAOYSA-N
Synonym 2-trimethylacetyl thiophene, 2,2-dimethyl-1-thiophen-2-yl propan-1-one, 2,2-dimethyl-1-2-thienyl propan-1-one, 2-pivaloylthiophene, t-butyl-2-thienylketone, acmc-1cb4m, 2-2,2-dimethylpropionyl thiophene, 2,2-dimethyl-1-thiophenyl-1-propanone, 2.2-dimethyl-1-thiophenyl-1-propanone
PubChem CID 2776976
IUPAC Name 2,2-dimethyl-1-thiophen-2-ylpropan-1-one
SMILES CC(C)(C)C(=O)C1=CC=CS1
Specifications
Density 1.07
Boiling Point 114°C to 116°C (14 mmHg)
Flash Point 99°C (210°F)
Refractive Index 1.535
Quantity 5 g
Beilstein 116243
Formula Weight 168.26
Percent Purity 98%
Chemical Name or Material 2-(Trimethylacetyl)thiophene