missing translation for 'onlineSavingsMsg'
Learn More

2-(Trihexylammonium)ethyl Methanethiosulfonate Bromide, TRC

Product Code. 30456116
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456116 100 mg 100mg
30451592 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456116 Supplier Toronto Research Chemicals Supplier No. TRCT794550100MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Trihexylammonium)ethyl Methanethiosulfonate Bromide
Analyte or Component Names 2-(Trihexylammonium)ethyl Methanethiosulfonate Bromide
Shipping Condition Room Temperature
Molecular Formula C21H46BrNO2S2
Synonym MTS-THAE
InChI Formula InChI=1S/C21H46NO2S2.BrH/c1-5-8-11-14-17-22(18-15-12-9-6-2,19-16-13-10-7-3)20-21-25-26(4,23)24;/h5-21H2,1-4H3;1H/q+1;/p-1
Solution Type Neat
SMILES O=S(SCC[N+](CCCCCC)(CCCCCC)CCCCCC)(C)=O.[Br-]
Industry Type Pharmaceutical
Molecular Weight (g/mol) 488.63
Quantity 100 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.